Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014146.3 | NP_054865.2 | 5, 6 | 598 | ||
NM_032463.2 | NP_115852.1 | 6, 7 | 717 | |||
NM_032464.2 | NP_115853.2 | 6, 7 | 787 | |||
XM_011516558.2 | XP_011514860.1 | 6, 7 | 627 | |||
GenBank mRNA | AF252611.1 | - | 6, 7 | 538 | ||
AF252612.1 | - | 5, 6 | 349 | |||
AF252613.1 | - | 5, 6 | 334 | |||
AF252614.1 | - | 6, 7 | 468 | |||
AF257135.1 | - | 4, 5 | 269 | |||
AK002099.1 | - | 6, 7 | 577 | |||
AK092904.1 | - | 6, 7 | 928 | |||
AK290074.1 | - | 6, 7 | 465 | |||
AK290916.1 | - | 5, 6 | 346 | |||
AY190023.1 | - | 4, 5 | 279 | |||
BC001609.1 | - | 6, 7 | 599 | 3543310 | ||
BC009204.2 | - | 4, 5 | 271 | 4054137 |