Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256171.1 | NP_001243100.1 | 5 | 792 | ||
NM_001256173.1 | NP_001243102.1 | 3 | 575 | |||
NM_003429.4 | NP_003420.2 | 4 | 702 | |||
XM_011528263.2 | XP_011526565.1 | 6 | 863 | |||
XM_011528264.1 | XP_011526566.1 | 2 | 1023 | |||
XM_017027234.1 | XP_016882723.1 | 2 | 502 | |||
GenBank mRNA | AK308258.1 | - | 4 | 532 | ||
BC047646.1 | - | 2 | 1005 | 4828290 | ||
DB119881.1 | - | 3 | 524 | |||
M61866.1 | - | 4 | 666 | |||
U35376.1 | - | 4 | 672 |