Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278510.2 | NP_001265439.2 | 6 | 2480 | ||
NM_014518.3 | NP_055333.3 | 6 | 2609 | |||
NR_103551.2 | 6 | 3064 | ||||
XM_006723372.3 | XP_006723435.1 | 6 | 2683 | |||
XM_011527292.1 | XP_011525594.1 | 6 | 2591 | |||
XM_017027280.1 | XP_016882769.1 | 6 | 3138 | |||
GenBank mRNA | AB209357.1 | - | 6 | 3033 | ||
AK091541.1 | - | 1 | 170 | |||
BC150612.1 | - | 4 | 2345 | 9056982 | ||
CR936724.1 | - | 6 | 2482 |