Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAX8 SIRNA Assays ›
Gene Symbol:
PAX8
Gene Name:
paired box 8
Gene Aliases:
-
Chromosome Location:
Chr.2: 113215997 - 113278950 on Build GRCh38
UniGene ID:
Hs.469728
Species:
Human
siRNA ID
HSS144580
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAX8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003466.3
NP_003457.1
12
1457
NM_013952.3
NP_039246.1
12
1378
NM_013953.3
NP_039247.1
10
1147
NM_013992.3
NP_054698.1
9
1045
XM_011511790.1
XP_011510092.1
12
1489
XM_011511791.1
XP_011510093.1
12
1545
XM_011511792.1
XP_011510094.1
12
1410
XM_011511793.1
XP_011510095.1
11
1300
XM_011511794.1
XP_011510096.1
11
1269
GenBank mRNA
AK096795.1
-
1
967
AK225538.1
-
12
1288
AK292191.1
-
12
1454
AK307173.1
-
4
295
BC001060.2
-
12
1448
2963877
CR936798.1
-
1
6942
KC733810.1
-
12
1554
KJ545852.1
-
2
300
KJ545853.1
-
2
300
KJ545855.1
-
2
300
KJ545856.1
-
2
299
KJ545857.1
-
2
300
KJ545858.1
-
2
300
KJ545859.1
-
2
300
KJ545861.1
-
2
324
KJ545862.1
-
2
300
KJ545863.1
-
2
300
KJ545867.1
-
2
301
KJ545868.1
-
2
300
KJ545869.1
-
2
318
KJ545870.1
-
2
300
KJ545871.1
-
2
300
KJ545873.1
-
2
299
KJ545875.1
-
2
318
KJ545877.1
-
2
318
KJ545883.1
-
2
300
KJ545884.1
-
2
300
KU178404.1
-
11
1291
KU178405.1
-
10
1102
KU178406.1
-
10
1093
L19606.1
-
11
1301
S77904.1
-
12
1372
S77905.1
-
10
1141
S77906.2
-
9
1039
X69699.1
-
12
1451
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
urogenital system development
branching involved in ureteric bud morphogenesis
kidney development
mesonephros development
mesenchymal to epithelial transition involved in metanephros morphogenesis
transcription, DNA-templated
transcription from RNA polymerase II promoter
sulfur compound metabolic process
central nervous system development
anatomical structure morphogenesis
thyroid gland development
thyroid-stimulating hormone signaling pathway
pronephric field specification
inner ear morphogenesis
regulation of apoptotic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
pronephros development
cellular response to gonadotropin stimulus
otic vesicle development
positive regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
metanephric epithelium development
metanephric distal convoluted tubule development
metanephric comma-shaped body morphogenesis
metanephric S-shaped body morphogenesis
metanephric nephron tubule formation
negative regulation of mesenchymal cell apoptotic process involved in metanephric nephron morphogenesis
regulation of metanephric nephron tubule epithelial cell differentiation
positive regulation of branching involved in ureteric bud morphogenesis
negative regulation of mesenchymal cell apoptotic process involved in metanephros development
negative regulation of apoptotic process involved in metanephric collecting duct development
negative regulation of apoptotic process involved in metanephric nephron tubule development
positive regulation of metanephric DCT cell differentiation
positive regulation of thyroid hormone generation
regulation of thyroid-stimulating hormone secretion
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
thyroid-stimulating hormone receptor activity
protein binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0