Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST2H2BE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003528.2 | NP_003519.1 | 1 | 91 | ||
GenBank mRNA | AK093747.1 | - | 1 | 91 | ||
AK289725.1 | - | 2 | 449 | |||
BC005827.2 | - | 1 | 61 | 2989788 | ||
BC069193.1 | - | 1 | 73 | 3936695 | ||
BC096121.2 | - | 1 | 49 | 40002353 | ||
BC098112.1 | - | 1 | 49 | 40014245 | ||
BC098289.1 | - | 1 | 49 | 40014249 | ||
BC107084.1 | - | 1 | 50 | 40014260 | ||
BC107085.1 | - | 1 | 50 | 40014264 | ||
CR541895.1 | - | 1 | 49 |
Gene Symbol: HIST2H2BC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036461.1 | 1 | 49 | |||
GenBank mRNA | AK299891.1 | - | 1 | 213 |