Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF020833.1 | - | 6, 7 | 422 | ||
AF094850.1 | - | 6, 7 | 440 | |||
AK291320.1 | - | 6, 7 | 441 | |||
AK293374.1 | - | 6, 7 | 525 | |||
AK316040.1 | - | 5, 6 | 570 | |||
AK316335.1 | - | 5, 6 | 576 | |||
BC000733.2 | - | 6, 7 | 441 | 3504640 | ||
BC008469.1 | - | 6, 7 | 442 | 4279770 | ||
BF701069.1 | - | 6, 7 | 457 | |||
BT006889.1 | - | 6, 7 | 399 | |||
CR457134.1 | - | 6, 7 | 399 | |||
DC356593.1 | - | 6, 7 | 464 | |||
U96074.1 | - | 6, 7 | 408 |