Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CACNA1G SIRNA Assays ›
Gene Symbol:
CACNA1G
Gene Name:
calcium voltage-gated channel subunit alpha1 G
Gene Aliases:
Ca(V)T.1, Cav3.1, NBR13, SCA42
Chromosome Location:
Chr.17: 50560929 - 50629625 on Build GRCh38
UniGene ID:
Hs.591169
Species:
Human
siRNA ID
HSS144925
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CACNA1G
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256324.1
NP_001243253.1
8
2202
NM_001256325.1
NP_001243254.1
8
2202
NM_001256326.1
NP_001243255.1
8
2202
NM_001256327.1
NP_001243256.1
8
2202
NM_001256328.1
NP_001243257.1
8
2202
NM_001256329.1
NP_001243258.1
8
2202
NM_001256330.1
NP_001243259.1
8
2202
NM_001256331.1
NP_001243260.1
8
2202
NM_001256332.1
NP_001243261.1
8
2202
NM_001256333.1
NP_001243262.1
8
2202
NM_001256334.1
NP_001243263.1
8
2202
NM_001256359.1
NP_001243288.1
8
2202
NM_001256360.1
NP_001243289.1
8
2202
NM_001256361.1
NP_001243290.1
8
2202
NM_018896.4
NP_061496.2
8
2202
NM_198376.2
NP_938190.1
8
2202
NM_198377.2
NP_938191.2
8
2202
NM_198378.2
NP_938192.1
8
2202
NM_198379.2
NP_938193.1
8
2202
NM_198380.2
NP_938194.1
8
2202
NM_198382.2
NP_938196.1
8
2202
NM_198383.2
NP_938197.1
8
2202
NM_198384.2
NP_938198.1
8
2202
NM_198385.2
NP_938199.1
8
2202
NM_198386.2
NP_938200.1
8
2202
NM_198387.2
NP_938201.1
8
2202
NM_198388.2
NP_938202.1
8
2202
NM_198396.2
NP_938406.1
8
2202
NR_046054.1
8
2202
NR_046055.1
8
2202
NR_046056.1
8
2202
NR_046057.1
8
2202
NR_046058.1
8
2202
XM_006722160.3
XP_006722223.1
8
2341
XM_006722161.3
XP_006722224.1
8
2290
GenBank mRNA
AB012043.1
-
8
1838
AB032949.2
-
1
164
AF124351.1
-
8
2182
AF126965.1
-
8
1985
AF126966.1
-
8
1985
AF134985.1
-
8
1810
AF134986.1
-
8
1810
AF190860.1
-
8
2026
AF227744.1
-
8
1810
AF227745.1
-
8
1810
AF227746.1
-
8
1810
AF227747.1
-
8
1810
AF227748.1
-
8
1810
AF227749.1
-
8
1810
AF227750.1
-
8
1810
AF227751.1
-
8
1810
BC110995.1
-
8
2202
5502027
DQ494449.1
-
8
1810
DQ494450.1
-
8
1810
DQ494451.1
-
8
1810
DQ494452.1
-
8
1810
DQ494453.1
-
8
1810
DQ494454.1
-
8
1810
DQ494455.1
-
8
1810
DQ494456.1
-
8
1810
DQ494457.1
-
8
1810
DQ494458.1
-
8
1810
DQ494459.1
-
8
1810
DQ494460.1
-
8
1810
DQ494461.1
-
8
1810
DQ494462.1
-
8
1810
DQ494463.1
-
8
1810
DQ494464.1
-
8
1810
DQ494465.1
-
8
1810
DQ494466.1
-
8
1810
DQ494467.1
-
8
1810
DQ494468.1
-
8
1810
DQ494469.1
-
8
1810
DQ494470.1
-
8
1810
DQ494471.1
-
8
1810
DQ494472.1
-
8
1810
DQ494473.1
-
8
1810
DQ494474.1
-
8
1810
DQ494475.1
-
8
1810
DQ494476.1
-
8
1810
DQ494477.1
-
8
1810
DQ494478.1
-
8
1810
DQ494479.1
-
8
1810
Back To Top
More Information
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
response to nickel cation
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of calcium ion-dependent exocytosis
regulation of atrial cardiac muscle cell membrane depolarization
calcium ion import
calcium ion transmembrane transport
membrane depolarization during action potential
SA node cell action potential
AV node cell action potential
SA node cell to atrial cardiac muscle cell signalling
AV node cell to bundle of His cell signaling
membrane depolarization during AV node cell action potential
membrane depolarization during SA node cell action potential
regulation of heart rate by cardiac conduction
low voltage-gated calcium channel activity
voltage-gated calcium channel activity involved in AV node cell action potential
voltage-gated calcium channel activity involved SA node cell action potential
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0