Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CACNA1G SIRNA Assays ›
Gene Symbol:
CACNA1G
Gene Name:
calcium voltage-gated channel subunit alpha1 G
Gene Aliases:
Ca(V)T.1, Cav3.1, NBR13, SCA42
Chromosome Location:
Chr.17: 50560929 - 50629625 on Build GRCh38
UniGene ID:
Hs.591169
Species:
Human
siRNA ID
HSS144926
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CACNA1G
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256324.1
NP_001243253.1
12
3094
NM_001256325.1
NP_001243254.1
12
3094
NM_001256326.1
NP_001243255.1
12
3094
NM_001256327.1
NP_001243256.1
12
3094
NM_001256328.1
NP_001243257.1
12
3094
NM_001256329.1
NP_001243258.1
12
3094
NM_001256330.1
NP_001243259.1
12
3094
NM_001256331.1
NP_001243260.1
12
3094
NM_001256332.1
NP_001243261.1
12
3094
NM_001256333.1
NP_001243262.1
12
3094
NM_001256334.1
NP_001243263.1
12
3094
NM_001256359.1
NP_001243288.1
12
3094
NM_001256360.1
NP_001243289.1
12
3094
NM_001256361.1
NP_001243290.1
12
3094
NM_018896.4
NP_061496.2
12
3094
NM_198376.2
NP_938190.1
12
3094
NM_198377.2
NP_938191.2
12
3094
NM_198378.2
NP_938192.1
12
3094
NM_198379.2
NP_938193.1
12
3094
NM_198380.2
NP_938194.1
12
3094
NM_198382.2
NP_938196.1
12
3094
NM_198383.2
NP_938197.1
12
3094
NM_198384.2
NP_938198.1
12
3094
NM_198385.2
NP_938199.1
12
3094
NM_198386.2
NP_938200.1
12
3094
NM_198387.2
NP_938201.1
12
3094
NM_198388.2
NP_938202.1
12
3094
NM_198396.2
NP_938406.1
12
3094
NR_046054.1
12
3094
NR_046055.1
12
3094
NR_046056.1
12
3094
NR_046057.1
12
3094
NR_046058.1
12
3094
XM_006722160.3
XP_006722223.1
12
3233
XM_006722161.3
XP_006722224.1
12
3182
GenBank mRNA
AB012043.1
-
12
2730
AB032949.2
-
5
1056
AF124351.1
-
12
3074
AF126965.1
-
12
2877
AF126966.1
-
12
2877
AF134985.1
-
12
2702
AF134986.1
-
12
2702
AF190860.1
-
12
2918
AF227744.1
-
12
2702
AF227745.1
-
12
2702
AF227746.1
-
12
2702
AF227747.1
-
12
2702
AF227748.1
-
12
2702
AF227749.1
-
12
2702
AF227750.1
-
12
2702
AF227751.1
-
12
2702
BC110995.1
-
12
3094
5502027
DQ494449.1
-
12
2702
DQ494450.1
-
12
2702
DQ494451.1
-
12
2702
DQ494452.1
-
12
2702
DQ494453.1
-
12
2702
DQ494454.1
-
12
2702
DQ494455.1
-
12
2702
DQ494456.1
-
12
2702
DQ494457.1
-
12
2702
DQ494458.1
-
12
2702
DQ494459.1
-
12
2702
DQ494460.1
-
12
2702
DQ494461.1
-
12
2702
DQ494462.1
-
12
2702
DQ494463.1
-
12
2702
DQ494464.1
-
12
2702
DQ494465.1
-
12
2702
DQ494466.1
-
12
2702
DQ494467.1
-
12
2702
DQ494468.1
-
12
2702
DQ494469.1
-
12
2702
DQ494470.1
-
12
2702
DQ494471.1
-
12
2702
DQ494472.1
-
12
2702
DQ494473.1
-
12
2702
DQ494474.1
-
12
2702
DQ494475.1
-
12
2702
DQ494476.1
-
12
2702
DQ494477.1
-
12
2702
DQ494478.1
-
12
2702
DQ494479.1
-
12
2702
Back To Top
More Information
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
response to nickel cation
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of calcium ion-dependent exocytosis
regulation of atrial cardiac muscle cell membrane depolarization
calcium ion import
calcium ion transmembrane transport
membrane depolarization during action potential
SA node cell action potential
AV node cell action potential
SA node cell to atrial cardiac muscle cell signalling
AV node cell to bundle of His cell signaling
membrane depolarization during AV node cell action potential
membrane depolarization during SA node cell action potential
regulation of heart rate by cardiac conduction
low voltage-gated calcium channel activity
voltage-gated calcium channel activity involved in AV node cell action potential
voltage-gated calcium channel activity involved SA node cell action potential
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0