Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317800.1 | NP_001304729.1 | 3 | 938 | ||
NM_003974.3 | NP_003965.2 | 5 | 1308 | |||
NM_201349.2 | NP_958728.1 | 4 | 1220 | |||
XM_005273680.4 | XP_005273737.1 | 4 | 1035 | |||
GenBank mRNA | AF034970.1 | - | 5 | 1201 | ||
AK226110.1 | - | 3 | 908 | |||
AK292126.1 | - | 5 | 1264 | |||
AK296671.1 | - | 4 | 1202 | |||
AL545478.3 | - | 1 | 464 | |||
BC032623.1 | - | 5 | 1276 | 5242017 |