Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005148.3 | NP_005139.1 | 5 | 750 | ||
NM_054035.2 | NP_473376.1 | 4 | 1017 | |||
XM_017025289.1 | XP_016880778.1 | 6 | 775 | |||
GenBank mRNA | AB593014.1 | - | 4 | 1020 | ||
AF028788.1 | - | 5 | 682 | |||
AF028789.1 | - | 4 | 950 | |||
AK124531.1 | - | 4 | 3556 | |||
BC027176.1 | - | 5 | 724 | 4341426 | ||
BX641031.1 | - | 4 | 3479 | |||
BX647747.1 | - | 2 | 582 | |||
BX648530.1 | - | 3 | 1420 | |||
U40998.1 | - | 5 | 733 |