Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042422.2 | NP_001035887.1 | 4 | 828 | ||
NM_001042423.2 | NP_001035888.1 | 4 | 832 | |||
NM_001206950.1 | NP_001193879.1 | 4 | 844 | |||
NM_001206951.1 | NP_001193880.1 | 4 | 806 | |||
NM_001206952.1 | NP_001193881.1 | 4 | 773 | |||
NM_004207.3 | NP_004198.1 | 4 | 800 | |||
XM_011523607.1 | XP_011521909.1 | 4 | 2721 | |||
GenBank mRNA | AK127319.1 | - | 3 | 2988 | ||
AK223040.1 | - | 4 | 758 | |||
BC112267.1 | - | 3 | 719 | 8327735 | ||
BC112269.1 | - | 3 | 719 | 8327737 | ||
U81800.1 | - | 4 | 740 |