Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004214.4 | NP_004205.2 | 4 | 592 | ||
NM_198897.1 | NP_942600.1 | 4 | 592 | |||
GenBank mRNA | AF010187.2 | - | 4 | 489 | ||
AF171944.1 | - | 4 | 471 | |||
AF171945.1 | - | 4 | 471 | |||
AF171946.1 | - | 4 | 471 | |||
AF250391.1 | - | 4 | 471 | |||
AK223438.1 | - | 4 | 514 | |||
AK289562.1 | - | 4 | 514 | |||
AK296571.1 | - | 5 | 493 | |||
AK309974.1 | - | 4 | 514 | |||
BC014388.2 | - | 4 | 532 | 3504921 | ||
BC017448.1 | - | 4 | 488 | 3509570 | ||
CD109589.1 | - | 4 | 619 | |||
CR456749.1 | - | 4 | 471 |