Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004223.4 | NP_004214.1 | 4 | 478 | ||
NM_198183.2 | NP_937826.1 | 4 | 837 | |||
GenBank mRNA | AF031141.1 | - | 4 | 379 | ||
AF061736.1 | - | 4 | 429 | |||
AK093462.1 | - | 4 | 791 | |||
AK129621.1 | - | 4 | 349 | |||
AK226165.1 | - | 4 | 532 | |||
AK315755.1 | - | 4 | 397 | |||
BC032491.1 | - | 4 | 478 | 5246109 | ||
CR457126.1 | - | 4 | 382 |