Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ITGB1BP1 SIRNA Assays ›
Gene Symbol:
ITGB1BP1
Gene Name:
integrin subunit beta 1 binding protein 1
Gene Aliases:
ICAP-1A, ICAP-1B, ICAP-1alpha, ICAP1, ICAP1A, ICAP1B
Chromosome Location:
Chr.2: 9405681 - 9423569 on Build GRCh38
UniGene ID:
Hs.467662
Species:
Human
siRNA ID
HSS145082
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ITGB1BP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001319066.1
NP_001305995.1
3
474
NM_001319067.1
NP_001305996.1
3
376
NM_001319068.1
NP_001305997.1
3
347
NM_004763.4
NP_004754.1
3
362
NM_022334.4
NP_071729.1
3
362
NR_134951.1
3
351
NR_134952.1
3
362
NR_134953.1
2
255
NR_134954.1
3
362
NR_134955.1
3
362
NR_134957.1
2
255
XM_005246183.4
XP_005246240.1
4
1048
XM_005246184.4
XP_005246241.1
4
487
XM_005246185.4
XP_005246242.1
4
360
XM_005246189.4
XP_005246246.1
4
1013
XM_006711903.3
XP_006711966.1
3
592
XM_011510416.2
XP_011508718.1
4
682
XM_017005267.1
XP_016860756.1
4
2177
XM_017005268.1
XP_016860757.1
4
352
XM_017005269.1
XP_016860758.1
3
903
XM_017005270.1
XP_016860759.1
4
2186
GenBank mRNA
AF012023.1
-
3
298
AF012024.1
-
3
298
AI144519.1
-
3
388
AI336197.1
-
3
290
AK092115.1
-
249
AK093179.1
-
3
936
AK289817.1
-
3
275
BC012264.1
-
3
254
3863540
BE146261.1
-
1
36
BG032225.1
-
3
255
BM823938.1
-
2
82
BQ421551.1
-
3
246
BQ425555.1
-
3
307
BU520903.1
-
3
321
CR457006.1
-
2
130
CT002942.1
-
2
163
DA087859.1
-
3
250
DA751320.1
-
2
139
DB167886.1
-
3
257
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel endothelial cell proliferation involved in sprouting angiogenesis
transcription, DNA-templated
negative regulation of protein kinase activity
negative regulation of cell adhesion involved in substrate-bound cell migration
cell-matrix adhesion
Notch signaling pathway
integrin-mediated signaling pathway
positive regulation of cell proliferation
negative regulation of cell proliferation
positive regulation of endothelial cell migration
negative regulation of fibroblast migration
protein transport
cell migration
cell differentiation
biomineral tissue development
negative regulation of protein binding
activation of protein kinase B activity
integrin activation
regulation of cell adhesion mediated by integrin
tube formation
intracellular signal transduction
cellular response to vascular endothelial growth factor stimulus
regulation of GTPase activity
receptor clustering
cellular response to fibroblast growth factor stimulus
positive regulation of Notch signaling pathway
positive regulation of transcription from RNA polymerase II promoter
regulation of blood vessel size
myoblast migration
positive regulation of stress fiber assembly
positive regulation of cell division
positive regulation of focal adhesion assembly
negative regulation of focal adhesion assembly
positive regulation of protein kinase B signaling
negative regulation of ERK1 and ERK2 cascade
negative regulation of cell migration involved in sprouting angiogenesis
positive regulation of protein targeting to membrane
negative regulation of protein targeting to membrane
negative regulation of substrate adhesion-dependent cell spreading
regulation of integrin-mediated signaling pathway
GDP-dissociation inhibitor activity
integrin binding
protein binding
protein transporter activity
protein kinase binding
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0