Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADGRG1 SIRNA Assays ›
Gene Symbol:
ADGRG1
Gene Name:
adhesion G protein-coupled receptor G1
Gene Aliases:
BFPP, BPPR, GPR56, TM7LN4, TM7XN1
Chromosome Location:
Chr.16: 57619535 - 57665039 on Build GRCh38
UniGene ID:
Hs.513633
Species:
Human
siRNA ID
HSS145091
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADGRG1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145770.2
NP_001139242.1
13
1886
NM_001145771.2
NP_001139243.1
13
2342
NM_001145772.2
NP_001139244.1
13
2350
NM_001145773.2
NP_001139245.1
12
2221
NM_001145774.2
NP_001139246.1
13
2123
NM_001290142.1
NP_001277071.1
12
1444
NM_001290143.1
NP_001277072.1
12
1793
NM_001290144.1
NP_001277073.1
12
1962
NM_005682.6
NP_005673.3
13
1904
NM_201524.3
NP_958932.1
13
2328
NM_201525.3
NP_958933.1
12
2206
XM_005256237.2
XP_005256294.1
12
1940
XM_005256238.2
XP_005256295.1
13
2279
XM_005256239.2
XP_005256296.1
13
2041
XM_005256240.4
XP_005256297.1
12
1762
XM_005256241.4
XP_005256298.1
12
1766
XM_005256242.4
XP_005256299.1
12
1823
XM_005256244.3
XP_005256301.1
12
1915
XM_005256245.2
XP_005256302.1
12
1966
XM_005256246.2
XP_005256303.1
13
2026
XM_005256247.2
XP_005256304.1
12
1925
XM_005256248.2
XP_005256305.1
12
1990
XM_005256249.3
XP_005256306.1
12
1781
XM_005256251.4
XP_005256308.1
12
1807
XM_005256252.2
XP_005256309.1
12
1751
XM_005256254.2
XP_005256311.1
12
1951
XM_005256255.2
XP_005256312.1
13
2246
XM_006721338.2
XP_006721401.1
13
2062
XM_006721339.3
XP_006721402.1
13
1869
XM_006721340.3
XP_006721403.1
13
2127
XM_006721341.2
XP_006721404.1
13
2084
XM_006721342.2
XP_006721405.1
13
2047
XM_006721343.3
XP_006721406.1
13
2112
XM_006721344.2
XP_006721407.1
13
2069
XM_006721345.3
XP_006721408.1
13
1838
XM_006721346.2
XP_006721409.1
12
1693
XM_006721347.2
XP_006721410.1
13
2261
XM_011523461.2
XP_011521763.1
13
2030
XM_011523462.2
XP_011521764.1
12
1845
XM_011523463.2
XP_011521765.1
13
1855
XM_011523464.2
XP_011521766.1
13
2015
XM_011523465.2
XP_011521767.1
13
1953
XM_011523466.2
XP_011521768.1
12
2000
XM_011523467.2
XP_011521769.1
13
1747
XM_011523468.2
XP_011521770.1
12
1749
XM_017023892.1
XP_016879381.1
12
1933
GenBank mRNA
AF106858.1
-
12
1774
AJ011001.1
-
13
1910
AK075270.1
-
13
1838
AK075555.1
-
13
1861
AK131550.1
-
12
1399
AK297236.1
-
12
1835
AK299110.1
-
12
1750
AK312405.1
-
12
2042
AY358400.1
-
13
1825
BC008770.2
-
12
1770
3139174
BC013207.1
-
13
1900
BT007311.1
-
11
1612
CR936747.1
-
12
1735
EU432119.1
-
11
1612
KU178472.1
-
11
1612
KU178473.1
-
11
1594
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
cell adhesion
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
Rho protein signal transduction
cell-cell signaling
brain development
negative regulation of cell proliferation
vascular endothelial growth factor production
cell migration
cerebral cortex regionalization
cerebral cortex radial glia guided migration
layer formation in cerebral cortex
positive regulation of Rho protein signal transduction
positive regulation of cell adhesion
protein kinase C signaling
positive regulation of neural precursor cell proliferation
negative regulation of neuron migration
G-protein coupled receptor activity
collagen binding
heparin binding
extracellular matrix binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline