Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201362.1 | NP_001188291.1 | 1, 2 | 191 | ||
NM_001201363.1 | NP_001188292.1 | 1, 2 | 191 | |||
NM_001318884.1 | NP_001305813.1 | 1, 2 | 191 | |||
NM_001318886.1 | NP_001305815.1 | 1, 2 | 191 | |||
NM_001318887.1 | NP_001305816.1 | 1, 2 | 191 | |||
NM_001318888.1 | NP_001305817.1 | 1, 2 | 191 | |||
NM_004242.3 | NP_004233.1 | 1, 2 | 191 | |||
NM_138730.2 | NP_620058.1 | 1, 2 | 191 | |||
GenBank mRNA | AF274949.1 | - | 1, 2 | 110 | ||
AF401520.1 | - | 1, 2 | 82 | |||
AK315391.1 | - | 1, 2 | 135 | |||
AY043282.1 | - | 1, 2 | 158 | |||
BC009529.2 | - | 1, 2 | 119 | 3890883 | ||
BF790582.1 | - | 1, 2 | 87 | |||
BI551210.1 | - | 1, 2 | 198 | |||
BM927225.1 | - | 1, 2 | 67 | |||
CF553005.1 | - | 1, 2 | 55 | |||
CK023734.1 | - | 1, 2 | 129 | |||
CX761444.1 | - | 1, 2 | 97 | |||
DA894213.1 | - | 1, 2 | 135 | |||
L40357.1 | - | 1, 2 | 76 |