Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN3 SIRNA Assays ›
Gene Symbol:
NRXN3
Gene Name:
neurexin 3
Gene Aliases:
C14orf60
Chromosome Location:
Chr.14: 77979904 - 79868290 on Build GRCh38
UniGene ID:
Hs.368307
Species:
Human
siRNA ID
HSS145127
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105250.2
NP_001098720.1
5
1821
NM_001272020.1
NP_001258949.1
4
1731
NM_004796.5
NP_004787.2
15
3184
NM_138970.4
NP_620426.2
4
1731
NR_073546.1
5
1821
NR_073547.1
18
4855
XM_005268218.2
XP_005268275.1
21
4924
XM_006720322.2
XP_006720385.1
20
4912
XM_006720323.2
XP_006720386.1
19
4888
XM_011537363.1
XP_011535665.1
21
4924
XM_011537364.2
XP_011535666.1
22
4886
XM_011537365.2
XP_011535667.1
21
4712
XM_011537366.1
XP_011535668.1
20
4900
XM_011537367.1
XP_011535669.1
20
4834
XM_011537368.1
XP_011535670.1
21
4924
XM_011537369.1
XP_011535671.1
21
4924
XM_011537370.1
XP_011535672.1
20
4900
XM_011537371.1
XP_011535673.1
21
4924
XM_011537372.1
XP_011535674.1
21
4924
XM_011537373.1
XP_011535675.1
20
4834
XM_011537377.2
XP_011535679.1
17
3216
XM_017021790.1
XP_016877279.1
22
5024
XM_017021791.1
XP_016877280.1
22
4807
XM_017021792.1
XP_016877281.1
20
4707
XM_017021793.1
XP_016877282.1
20
4897
XM_017021794.1
XP_016877283.1
19
4888
XM_017021795.1
XP_016877284.1
19
4683
XM_017021796.1
XP_016877285.1
19
4873
XM_017021797.1
XP_016877286.1
18
4864
XM_017021798.1
XP_016877287.1
18
4861
XM_017021799.1
XP_016877288.1
20
4834
XM_017021800.1
XP_016877289.1
20
4834
XM_017021801.1
XP_016877290.1
19
4807
XM_017021802.1
XP_016877291.1
18
4828
XM_017021803.1
XP_016877292.1
20
4903
XM_017021804.1
XP_016877293.1
20
4897
XM_017021805.1
XP_016877294.1
19
4888
XM_017021806.1
XP_016877295.1
19
4873
XM_017021807.1
XP_016877296.1
20
4834
GenBank mRNA
AB018286.1
-
15
3165
AJ316284.2
-
18
4643
AJ493127.1
-
5
897
AK056530.1
-
4
1731
AL833739.1
-
4
1026
BC059368.1
-
4
1657
BC068469.1
-
4
1657
BC142649.1
-
5
1254
BC150194.1
-
5
1254
BC152457.1
-
15
3165
8862590
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
neurotransmitter secretion
axon guidance
synapse assembly
learning
adult behavior
social behavior
positive regulation of synapse assembly
vocalization behavior
positive regulation of synapse maturation
receptor activity
calcium channel regulator activity
metal ion binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0