Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN1 SIRNA Assays ›
Gene Symbol:
NRXN1
Gene Name:
neurexin 1
Gene Aliases:
Hs.22998, PTHSL2, SCZD17
Chromosome Location:
Chr.2: 49918503 - 51032536 on Build GRCh38
UniGene ID:
Hs.637685
Species:
Human
siRNA ID
HSS145133
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135659.1
NP_001129131.1
24
5849
NM_001320156.1
NP_001307085.1
3
360
NM_001320157.1
NP_001307086.1
3
351
NM_004801.4
NP_004792.1
22
5639
NM_138735.2
NP_620072.1
6
1864
XM_005264642.3
XP_005264699.1
23
5741
XM_005264643.3
XP_005264700.1
22
5696
XM_006712137.3
XP_006712200.1
22
5651
XM_011533167.2
XP_011531469.1
23
5750
XM_011533171.2
XP_011531473.1
23
5729
XM_011533172.2
XP_011531474.1
22
5723
XM_011533174.2
XP_011531476.1
22
5705
XM_011533175.2
XP_011531477.1
21
5693
XM_011533177.2
XP_011531479.1
22
5660
XM_011533178.2
XP_011531480.1
20
5660
XM_011533183.1
XP_011531485.1
17
3432
XM_017005303.1
XP_016860792.1
24
5777
XM_017005304.1
XP_016860793.1
24
5774
XM_017005305.1
XP_016860794.1
24
5768
XM_017005306.1
XP_016860795.1
23
5765
XM_017005307.1
XP_016860796.1
23
5759
XM_017005308.1
XP_016860797.1
24
5756
XM_017005309.1
XP_016860798.1
23
5750
XM_017005310.1
XP_016860799.1
22
5747
XM_017005311.1
XP_016860800.1
23
5732
XM_017005312.1
XP_016860801.1
23
5726
XM_017005313.1
XP_016860802.1
23
5720
XM_017005314.1
XP_016860803.1
21
5717
XM_017005315.1
XP_016860804.1
22
5714
XM_017005316.1
XP_016860805.1
22
5714
XM_017005317.1
XP_016860806.1
22
5708
XM_017005318.1
XP_016860807.1
22
5705
XM_017005319.1
XP_016860808.1
22
5702
XM_017005320.1
XP_016860809.1
22
5693
XM_017005321.1
XP_016860810.1
23
5687
XM_017005322.1
XP_016860811.1
23
5678
XM_017005323.1
XP_016860812.1
19
5663
XM_017005324.1
XP_016860813.1
21
5633
XM_017005325.1
XP_016860814.1
21
5624
XM_017005326.1
XP_016860815.1
21
5621
XM_017005327.1
XP_016860816.1
21
5606
XM_017005328.1
XP_016860817.1
20
5588
XM_017005330.1
XP_016860819.1
18
5564
XM_017005331.1
XP_016860820.1
18
5549
XM_017005332.1
XP_016860821.1
17
5528
XM_017005333.1
XP_016860822.1
17
5519
XM_017005334.1
XP_016860823.1
20
3598
XM_017005335.1
XP_016860824.1
7
6944
XM_017005336.1
XP_016860825.1
7
6936
XM_017005337.1
XP_016860826.1
6
6845
GenBank mRNA
AB011150.2
-
21
4402
AB035356.1
-
21
4354
AF064842.1
-
6
1863
AK295773.1
-
3
254
BC125179.1
-
7
920
BC125180.1
-
7
1283
BC150247.1
-
21
4423
9007472
BX647616.1
-
3
314
DA322531.1
-
3
263
DC324848.1
-
3
360
DC333548.1
-
3
323
EF539882.1
-
24
5712
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
chemical synaptic transmission
neurotransmitter secretion
axon guidance
synapse assembly
learning
adult behavior
social behavior
vocal learning
neuromuscular process controlling balance
positive regulation of synapse assembly
positive regulation of synaptic transmission, glutamatergic
prepulse inhibition
regulation of insulin secretion involved in cellular response to glucose stimulus
vocalization behavior
positive regulation of synapse maturation
postsynaptic membrane assembly
gephyrin clustering involved in postsynaptic density assembly
neuroligin clustering involved in postsynaptic membrane assembly
postsynaptic density protein 95 clustering
positive regulation of excitatory postsynaptic potential
regulation of grooming behavior
receptor activity
calcium channel regulator activity
calcium ion binding
protein binding
acetylcholine receptor binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline