Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBM39 SIRNA Assays ›
Gene Symbol:
RBM39
Gene Name:
RNA binding motif protein 39
Gene Aliases:
CAPER, CAPERalpha, FSAP59, HCC1, RNPC2
Chromosome Location:
Chr.20: 35703609 - 35742336 on Build GRCh38
UniGene ID:
Hs.282901
Species:
Human
siRNA ID
HSS145212
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBM39
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242599.1
NP_001229528.1
4
587
NM_001242600.1
NP_001229529.1
4
587
NM_001323422.1
NP_001310351.1
4
587
NM_001323423.1
NP_001310352.1
6
732
NM_001323424.1
NP_001310353.1
4
587
NM_004902.3
NP_004893.1
4
587
NM_184234.2
NP_909122.1
4
587
NR_040722.1
5
660
NR_040723.1
5
659
NR_040724.1
5
660
XM_006723890.3
XP_006723953.1
4
2167
XM_006723891.3
XP_006723954.1
5
1190
XM_011529111.2
XP_011527413.1
5
2239
XM_017028138.1
XP_016883627.1
4
587
XM_017028139.1
XP_016883628.1
6
1556
XM_017028140.1
XP_016883629.1
5
660
XM_017028141.1
XP_016883630.1
4
2167
XM_017028142.1
XP_016883631.1
5
3063
XM_017028143.1
XP_016883632.1
5
2239
XM_017028144.1
XP_016883633.1
6
732
XM_017028145.1
XP_016883634.1
5
660
XM_017028146.1
XP_016883635.1
5
660
XM_017028147.1
XP_016883636.1
5
583
GenBank mRNA
AK126158.1
-
1
746
AK299165.1
-
4
487
AK299678.1
-
4
489
AK303910.1
-
4
459
AL833168.1
-
4
473
BC107886.1
-
5
369
BC131543.1
-
4
353
40085367
BC141835.1
-
4
437
40148871
BC158172.1
-
4
437
40148190
BX537770.1
-
5
579
BX640714.1
-
5
572
BX640812.1
-
5
569
CR749443.1
-
4
2254
DQ656052.2
-
4
194
L10910.1
-
4
327
L10911.1
-
3
202
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
RNA processing
mRNA processing
RNA splicing
nucleotide binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-k9xzd:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0