Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005103.4 | NP_005094.1 | 2 | 447 | ||
NM_022549.3 | NP_072043.1 | 2 | 447 | |||
XM_005271734.2 | XP_005271791.1 | 2 | 492 | |||
XM_005271735.2 | XP_005271792.1 | 2 | 544 | |||
GenBank mRNA | AK296554.1 | - | 2 | 354 | ||
BC009545.1 | - | 2 | 364 | 3897353 | ||
CN310685.1 | - | 2 | 389 | |||
CR456817.1 | - | 1 | 212 | |||
CR541954.1 | - | 1 | 212 | |||
DA697171.1 | - | 2 | 454 | |||
U60060.1 | - | 2 | 311 | |||
U60062.1 | - | 1 | 241 | |||
U69139.1 | - | 1 | 237 |