Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039459.2 | NP_001034548.1 | 3 | 433 | ||
NM_001206480.2 | NP_001193409.1 | 18 | 2212 | |||
NM_001206482.1 | NP_001193411.1 | 18 | 2382 | |||
NM_014800.10 | NP_055615.8 | 18 | 2023 | |||
NM_130442.3 | NP_569709.1 | 3 | 579 | |||
XM_005249919.2 | XP_005249976.1 | 19 | 2576 | |||
XM_006715805.1 | XP_006715868.1 | 18 | 1868 | |||
XM_011515654.1 | XP_011513956.1 | 19 | 3261 | |||
XM_017012838.1 | XP_016868327.1 | 19 | 3446 | |||
XM_017012839.1 | XP_016868328.1 | 18 | 1810 | |||
GenBank mRNA | AF398885.1 | - | 17 | 1674 | ||
AK126565.1 | - | 6 | 1063 | |||
AK291784.1 | - | 18 | 2382 | |||
AK296253.1 | - | 15 | 1543 | |||
AK314349.1 | - | 18 | 1812 | |||
AL136787.1 | - | 18 | 1924 | |||
BC003051.1 | - | 3 | 429 | |||
BC064635.1 | - | 3 | 381 | |||
BC077074.1 | - | 3 | 430 | |||
BC114341.2 | - | 18 | 1761 | 40035461 | ||
BI600726.1 | - | 3 | 441 | |||
D87457.1 | - | 3 | 405 |