Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZC3H11A SIRNA Assays ›
Gene Symbol:
ZC3H11A
Gene Name:
zinc finger CCCH-type containing 11A
Gene Aliases:
ZC3HDC11A
Chromosome Location:
Chr.1: 203795537 - 203854128 on Build GRCh38
UniGene ID:
Hs.532399
Species:
Human
siRNA ID
HSS145344
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZC3H11A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001319238.1
NP_001306167.1
16
2737
NM_001319239.1
NP_001306168.1
16
2632
NM_014827.5
NP_055642.3
16
2308
XM_005245643.2
XP_005245700.1
16
2327
XM_005245644.3
XP_005245701.1
15
2011
XM_006711673.2
XP_006711736.1
16
2174
XM_011510199.1
XP_011508501.1
14
3379
XM_011510200.1
XP_011508502.1
15
2399
XM_011510201.1
XP_011508503.1
16
2585
XM_011510202.1
XP_011508504.1
14
2324
XM_017002956.1
XP_016858445.1
15
3633
XM_017002957.1
XP_016858446.1
15
3485
XM_017002958.1
XP_016858447.1
15
3472
XM_017002959.1
XP_016858448.1
16
2545
XM_017002960.1
XP_016858449.1
15
3633
XM_017002961.1
XP_016858450.1
14
3379
XM_017002962.1
XP_016858451.1
15
2399
XM_017002963.1
XP_016858452.1
16
2191
XM_017002964.1
XP_016858453.1
15
3545
XM_017002965.1
XP_016858454.1
14
2874
GenBank mRNA
AB014563.1
-
13
1819
AK095858.1
-
16
2605
AK296983.1
-
12
1666
AK310470.1
-
4
701
BC014268.2
-
16
2146
3844154
BC038513.2
-
16
2463
BX649148.1
-
13
5924
CR627439.1
-
15
3329
CR627446.1
-
13
5432
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA processing factor
Gene Ontology Categories:
Function(s)
Process(es)
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
poly(A)+ mRNA export from nucleus
mRNA 3'-end processing
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0