Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242925.1 | NP_001229854.1 | 2 | 383 | ||
NM_014858.3 | NP_055673.2 | 2 | 1047 | |||
XM_005245684.1 | XP_005245741.1 | 2 | 738 | |||
XM_005245685.4 | XP_005245742.1 | 2 | 369 | |||
XM_005245686.3 | XP_005245743.1 | 2 | 764 | |||
GenBank mRNA | AB007950.3 | - | 2 | 1047 | ||
AK131419.1 | - | 3 | 836 | |||
AK293734.1 | - | 2 | 383 | |||
BC131629.1 | - | 2 | 519 | 40118729 | ||
DC309246.1 | - | 2 | 381 |