Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCGB1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006551.3 | NP_006542.1 | 2 | 257 | ||
GenBank mRNA | AJ224172.1 | - | 2 | 283 | ||
AL708956.1 | - | 2 | 282 | |||
BC069290.1 | - | 2 | 251 | 7262705 | ||
BC104838.1 | - | 2 | 227 | 8143841 | ||
BC104840.1 | - | 2 | 226 | 8143843 |