Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GADD45G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006705.3 | NP_006696.1 | 3 | 329 | ||
XM_011518163.2 | XP_011516465.1 | 2 | 230 | |||
GenBank mRNA | AF078078.1 | - | 3 | 234 | ||
AF079806.1 | - | 3 | 316 | |||
AF087883.1 | - | 3 | 314 | |||
AK313689.1 | - | 3 | 327 | |||
BC000465.2 | - | 3 | 310 | 2822765 | ||
BC019325.1 | - | 3 | 310 | 2823131 | ||
BT007234.1 | - | 3 | 219 | |||
CR457038.1 | - | 3 | 219 | |||
D83023.1 | - | 3 | 302 | |||
DA420966.1 | - | 3 | 329 |