Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014267.5 | NP_055082.1 | 1 | 194 | ||
GenBank mRNA | AK315382.1 | - | 1 | 148 | ||
BC007103.1 | - | 1 | 148 | 4281282 | ||
BC016352.1 | - | 1 | 152 | 4082845 | ||
BC020937.1 | - | 1 | 148 | 4281930 | ||
BC103747.1 | - | 1 | 175 | 6151644 | ||
BP210918.1 | - | 1 | 148 | |||
BP226882.1 | - | 1 | 394 | |||
BU661194.1 | - | 1 | 159 | |||
CR456882.1 | - | 1 | 16 | |||
U51678.1 | - | 1 | 51 |