Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NT5C2 SIRNA Assays ›
Gene Symbol:
NT5C2
Gene Name:
5'-nucleotidase, cytosolic II
Gene Aliases:
GMP, NT5B, PNT5, SPG45, SPG65, cN-II
Chromosome Location:
Chr.10: 103088017 - 103193306 on Build GRCh38
UniGene ID:
Hs.97439
Species:
Human
siRNA ID
HSS146101
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NT5C2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134373.2
NP_001127845.1
18
1665
NM_012229.4
NP_036361.1
19
1763
XM_005269632.4
XP_005269689.1
19
1674
XM_005269633.4
XP_005269690.1
20
1767
XM_005269634.4
XP_005269691.1
20
1799
XM_005269635.4
XP_005269692.1
19
1989
XM_005269636.4
XP_005269693.1
19
1775
XM_005269637.4
XP_005269694.1
18
2493
XM_005269638.4
XP_005269695.1
18
3794
XM_005269639.4
XP_005269696.1
17
1518
XM_005269640.4
XP_005269697.1
20
1790
XM_005269641.4
XP_005269698.1
19
1692
XM_005269642.4
XP_005269699.1
18
1617
XM_005269643.4
XP_005269700.1
17
1557
XM_005269644.4
XP_005269701.1
21
1814
XM_005269645.4
XP_005269702.1
20
1716
XM_005269646.4
XP_005269703.1
19
1642
XM_006717721.3
XP_006717784.1
19
1716
XM_006717723.3
XP_006717786.1
17
1513
XM_011539537.2
XP_011537839.1
19
1628
XM_017015947.1
XP_016871436.1
17
1543
XM_017015948.1
XP_016871437.1
20
1899
XM_017015949.1
XP_016871438.1
20
1884
XM_017015950.1
XP_016871439.1
18
1600
XM_017015951.1
XP_016871440.1
17
1526
XM_017015952.1
XP_016871441.1
19
1823
XM_017015953.1
XP_016871442.1
20
2218
XM_017015954.1
XP_016871443.1
19
2157
XM_017015955.1
XP_016871444.1
19
1844
XM_017015956.1
XP_016871445.1
19
2050
XM_017015957.1
XP_016871446.1
21
1908
XM_017015958.1
XP_016871447.1
22
2027
XM_017015959.1
XP_016871448.1
18
1618
XM_017015960.1
XP_016871449.1
19
1581
XM_017015961.1
XP_016871450.1
19
1976
XM_017015962.1
XP_016871451.1
19
1810
XM_017015963.1
XP_016871452.1
19
1914
XM_017015964.1
XP_016871453.1
19
1893
XM_017015965.1
XP_016871454.1
18
1770
XM_017015966.1
XP_016871455.1
18
1770
XM_017015967.1
XP_016871456.1
18
1705
XM_017015968.1
XP_016871457.1
19
1698
XM_017015969.1
XP_016871458.1
19
1792
XM_017015970.1
XP_016871459.1
17
2435
XM_017015971.1
XP_016871460.1
16
1479
XM_017015972.1
XP_016871461.1
20
1733
XM_017015973.1
XP_016871462.1
21
1757
XM_017015974.1
XP_016871463.1
20
1757
XM_017015975.1
XP_016871464.1
18
1598
XM_017015976.1
XP_016871465.1
18
1604
GenBank mRNA
AK127670.1
-
19
1754
AK291667.1
-
18
1631
AK295593.1
-
17
1807
AK312517.1
-
18
1631
BC001595.1
-
18
1714
3543103
BX648078.1
-
18
5665
D38524.1
-
18
1653
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
purine nucleotide catabolic process
phosphorylation
dephosphorylation
drug metabolic process
IMP metabolic process
adenosine metabolic process
nucleotide binding
protein binding
5'-nucleotidase activity
metal ion binding
nucleoside phosphotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0