Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NT5C2 SIRNA Assays ›
Gene Symbol:
NT5C2
Gene Name:
5'-nucleotidase, cytosolic II
Gene Aliases:
GMP, NT5B, PNT5, SPG45, SPG65, cN-II
Chromosome Location:
Chr.10: 103088017 - 103193306 on Build GRCh38
UniGene ID:
Hs.97439
Species:
Human
siRNA ID
HSS146103
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NT5C2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134373.2
NP_001127845.1
6
531
NM_012229.4
NP_036361.1
7
629
XM_005269632.4
XP_005269689.1
7
540
XM_005269633.4
XP_005269690.1
8
633
XM_005269634.4
XP_005269691.1
8
665
XM_005269635.4
XP_005269692.1
7
855
XM_005269636.4
XP_005269693.1
7
641
XM_005269637.4
XP_005269694.1
6
1359
XM_005269638.4
XP_005269695.1
6
2660
XM_005269639.4
XP_005269696.1
5
384
XM_005269640.4
XP_005269697.1
8
656
XM_005269641.4
XP_005269698.1
7
558
XM_005269642.4
XP_005269699.1
6
483
XM_005269643.4
XP_005269700.1
5
423
XM_005269644.4
XP_005269701.1
9
680
XM_005269645.4
XP_005269702.1
8
582
XM_005269646.4
XP_005269703.1
7
508
XM_006717721.3
XP_006717784.1
7
582
XM_006717723.3
XP_006717786.1
5
379
XM_011539537.2
XP_011537839.1
8
665
XM_017015947.1
XP_016871436.1
5
409
XM_017015948.1
XP_016871437.1
8
765
XM_017015949.1
XP_016871438.1
7
558
XM_017015952.1
XP_016871441.1
6
497
XM_017015953.1
XP_016871442.1
7
892
XM_017015954.1
XP_016871443.1
6
831
XM_017015955.1
XP_016871444.1
6
497
XM_017015956.1
XP_016871445.1
6
831
XM_017015957.1
XP_016871446.1
8
582
XM_017015958.1
XP_016871447.1
9
680
XM_017015959.1
XP_016871448.1
6
484
XM_017015960.1
XP_016871449.1
7
447
XM_017015961.1
XP_016871450.1
7
842
XM_017015962.1
XP_016871451.1
6
484
XM_017015963.1
XP_016871452.1
6
567
XM_017015964.1
XP_016871453.1
6
567
XM_017015965.1
XP_016871454.1
5
444
XM_017015966.1
XP_016871455.1
5
423
XM_017015967.1
XP_016871456.1
5
379
XM_017015972.1
XP_016871461.1
7
558
XM_017015973.1
XP_016871462.1
8
582
XM_017015974.1
XP_016871463.1
7
582
XM_017015975.1
XP_016871464.1
5
423
GenBank mRNA
AK127670.1
-
6
339
AK291667.1
-
6
497
AK295593.1
-
5
673
AK312517.1
-
6
497
BC001595.1
-
6
580
3543103
BX648078.1
-
8
662
D38524.1
-
6
519
DB169780.1
-
6
533
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
purine nucleotide catabolic process
phosphorylation
dephosphorylation
drug metabolic process
IMP metabolic process
adenosine metabolic process
nucleotide binding
protein binding
5'-nucleotidase activity
metal ion binding
nucleoside phosphotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0