Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLASP2 SIRNA Assays ›
Gene Symbol:
CLASP2
Gene Name:
cytoplasmic linker associated protein 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 33496245 - 33718356 on Build GRCh38
UniGene ID:
Hs.108614
Species:
Human
siRNA ID
HSS146201
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLASP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001207044.1
NP_001193973.1
32
3960
NM_015097.2
NP_055912.2
38
4382
XM_006713040.1
XP_006713103.1
40
4639
XM_006713041.1
XP_006713104.1
40
4636
XM_006713042.1
XP_006713105.1
39
4615
XM_006713048.1
XP_006713111.1
39
4576
XM_006713049.1
XP_006713112.1
38
4561
XM_006713050.1
XP_006713113.1
38
4552
XM_006713052.1
XP_006713115.1
38
4522
XM_006713053.1
XP_006713116.1
37
4501
XM_011533515.1
XP_011531817.1
38
4597
XM_017005947.1
XP_016861436.1
39
4612
XM_017005948.1
XP_016861437.1
38
4609
XM_017005949.1
XP_016861438.1
38
4440
XM_017005950.1
XP_016861439.1
39
4588
XM_017005951.1
XP_016861440.1
39
4585
XM_017005952.1
XP_016861441.1
38
4585
XM_017005953.1
XP_016861442.1
38
4582
XM_017005954.1
XP_016861443.1
39
4573
XM_017005955.1
XP_016861444.1
38
4564
XM_017005956.1
XP_016861445.1
37
4561
XM_017005957.1
XP_016861446.1
37
4549
XM_017005958.1
XP_016861447.1
37
4522
XM_017005959.1
XP_016861448.1
37
4519
XM_017005960.1
XP_016861449.1
36
4510
XM_017005961.1
XP_016861450.1
36
4353
XM_017005962.1
XP_016861451.1
37
4498
XM_017005963.1
XP_016861452.1
36
4498
XM_017005964.1
XP_016861453.1
36
4495
XM_017005965.1
XP_016861454.1
36
4483
XM_017005966.1
XP_016861455.1
35
4459
XM_017005967.1
XP_016861456.1
35
4302
XM_017005968.1
XP_016861457.1
33
3716
XM_017005969.1
XP_016861458.1
33
3726
XM_017005970.1
XP_016861459.1
33
3975
XM_017005971.1
XP_016861460.1
32
3702
XM_017005972.1
XP_016861461.1
33
4303
XM_017005973.1
XP_016861462.1
32
3948
XM_017005974.1
XP_016861463.1
31
3945
XM_017005975.1
XP_016861464.1
32
3665
XM_017005976.1
XP_016861465.1
30
3660
XM_017005977.1
XP_016861466.1
32
3924
XM_017005978.1
XP_016861467.1
31
3641
XM_017005979.1
XP_016861468.1
31
3651
XM_017005980.1
XP_016861469.1
32
4252
XM_017005981.1
XP_016861470.1
30
3638
XM_017005982.1
XP_016861471.1
30
3648
XM_017005983.1
XP_016861472.1
31
3900
XM_017005984.1
XP_016861473.1
31
3897
XM_017005985.1
XP_016861474.1
30
4225
XM_017005986.1
XP_016861475.1
29
3858
GenBank mRNA
AB014527.2
-
32
3599
AK024770.1
-
2
128
AK293963.1
-
4
385
AL137636.1
-
5
696
BC011530.1
-
18
2197
BC140778.1
-
38
4218
9021646
BX640725.1
-
14
1754
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor microtubule binding protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
vesicle targeting
microtubule nucleation
negative regulation of microtubule depolymerization
Golgi organization
mitotic spindle organization
sister chromatid cohesion
establishment or maintenance of cell polarity
exit from mitosis
regulation of gastrulation
positive regulation of epithelial cell migration
regulation of epithelial to mesenchymal transition
microtubule organizing center organization
regulation of microtubule polymerization or depolymerization
regulation of microtubule-based process
microtubule anchoring
positive regulation of exocytosis
cell division
negative regulation of stress fiber assembly
negative regulation of focal adhesion assembly
establishment of protein localization to plasma membrane
positive regulation of extracellular matrix disassembly
negative regulation of wound healing, spreading of epidermal cells
positive regulation of basement membrane assembly involved in embryonic body morphogenesis
dystroglycan binding
protein binding
microtubule binding
microtubule plus-end binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qvdz4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0