Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015135.2 | NP_055950.1 | 11 | 1544 | ||
XM_005250235.3 | XP_005250292.1 | 10 | 1496 | |||
GenBank mRNA | BC136624.1 | - | 11 | 1539 | 9020614 | |
BC146784.1 | - | 11 | 1518 | 9007240 | ||
D86978.1 | - | 11 | 1518 |