Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TBC1D12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015188.1 | NP_056003.1 | 13 | 2489 | ||
XM_006717732.3 | XP_006717795.2 | 12 | 2408 | |||
XM_006717733.2 | XP_006717796.1 | 14 | 1449 | |||
XM_011539558.2 | XP_011537860.1 | 13 | 1500 | |||
XM_017015996.1 | XP_016871485.1 | 11 | 2264 | |||
XM_017015998.1 | XP_016871487.1 | 12 | 1276 | |||
XM_017015999.1 | XP_016871488.1 | 12 | 1492 | |||
GenBank mRNA | AB011180.1 | - | 13 | 2305 | ||
AB449898.1 | - | 13 | 2286 | |||
AK090865.1 | - | 4 | 2117 | |||
AL049956.1 | - | 12 | 1237 | |||
BC064323.1 | - | 4 | 507 |