Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRUNE2 SIRNA Assays ›
Gene Symbol:
PRUNE2
Gene Name:
prune homolog 2
Gene Aliases:
BMCC1, BNIPXL, C9orf65, KIAA0367
Chromosome Location:
Chr.9: 76611376 - 76906220 on Build GRCh38
UniGene ID:
Hs.262857
Species:
Human
siRNA ID
HSS146296
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRUNE2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308047.1
NP_001294976.1
8
1373
NM_001308048.1
NP_001294977.1
8
1373
NM_015225.2
NP_056040.2
8
1373
NR_131751.1
7
1170
XM_005251745.1
XP_005251802.1
8
1506
XM_005251746.1
XP_005251803.1
8
1506
XM_005251748.1
XP_005251805.1
8
1506
XM_005251750.1
XP_005251807.1
8
1506
XM_005251751.1
XP_005251808.1
8
1506
XM_005251754.1
XP_005251811.1
8
1506
XM_006716982.1
XP_006717045.1
8
1506
XM_006716983.1
XP_006717046.1
8
1506
XM_006716984.2
XP_006717047.1
8
1506
XM_006716985.1
XP_006717048.1
8
1506
XM_006716986.1
XP_006717049.1
8
1506
XM_011518323.1
XP_011516625.1
8
1506
XM_011518326.2
XP_011516628.1
8
1506
XM_011518327.1
XP_011516629.1
8
1506
XM_011518328.2
XP_011516630.1
8
1506
XM_017014345.1
XP_016869834.1
8
1506
XM_017014346.1
XP_016869835.1
8
1506
XM_017014347.1
XP_016869836.1
8
1506
XM_017014348.1
XP_016869837.1
8
1506
XM_017014349.1
XP_016869838.1
8
1506
XM_017014350.1
XP_016869839.1
8
1506
XM_017014351.1
XP_016869840.1
8
1506
XM_017014352.1
XP_016869841.1
8
1506
XM_017014353.1
XP_016869842.1
8
1506
GenBank mRNA
AB050197.1
-
2
494
AB535152.1
-
8
1249
BC150641.1
-
2
391
9057011
FJ808772.1
-
8
1327
FJ808773.1
-
7
1124
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
pyrophosphatase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0