Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130963.1 | NP_001124435.1 | 1, 2 | 175 | ||
NM_015257.2 | NP_056072.2 | 1, 2 | 175 | |||
XM_011538059.1 | XP_011536361.1 | 1, 2 | 175 | |||
XM_017019079.1 | XP_016874568.1 | 1, 2 | 152 | |||
GenBank mRNA | AB006624.2 | - | 1, 2 | 136 | ||
AK294566.1 | - | 1, 2 | 147 | |||
AL833310.1 | - | 1, 2 | 153 | |||
AL833311.1 | - | 1, 2 | 153 | |||
BC071597.1 | - | 1, 2 | 147 | |||
BC117436.1 | - | 1, 2 | 147 | 40125987 | ||
BC126300.1 | - | 1, 2 | 147 | 8992016 | ||
CR749406.1 | - | 1, 2 | 147 | |||
DA724501.1 | - | 1, 2 | 175 | |||
DB123768.1 | - | 1, 2 | 175 |