Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130963.1 | NP_001124435.1 | 2, 3 | 295 | ||
NM_015257.2 | NP_056072.2 | 2, 3 | 295 | |||
XM_011538056.2 | XP_011536358.1 | 2, 3 | 975 | |||
XM_011538057.2 | XP_011536359.1 | 3, 4 | 397 | |||
XM_011538058.2 | XP_011536360.1 | 2, 3 | 972 | |||
XM_011538059.1 | XP_011536361.1 | 2, 3 | 295 | |||
XM_011538060.2 | XP_011536362.1 | 2, 3 | 1012 | |||
XM_017019077.1 | XP_016874566.1 | 2, 3 | 746 | |||
XM_017019078.1 | XP_016874567.1 | 2, 3 | 155 | |||
XM_017019079.1 | XP_016874568.1 | 2, 3 | 272 | |||
GenBank mRNA | AB006624.2 | - | 2, 3 | 256 | ||
AK294566.1 | - | 2, 3 | 267 | |||
AL833310.1 | - | 2, 3 | 273 | |||
AL833311.1 | - | 2, 3 | 273 | |||
BC071597.1 | - | 2, 3 | 267 | |||
BC117436.1 | - | 2, 3 | 267 | 40125987 | ||
BC126300.1 | - | 2, 3 | 267 | 8992016 | ||
CR749406.1 | - | 2, 3 | 267 | |||
DA724501.1 | - | 2, 3 | 295 | |||
DB123768.1 | - | 2, 3 | 295 |