Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESYT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184796.1 | NP_001171725.1 | 15, 16 | 1771 | ||
NM_015292.2 | NP_056107.1 | 15, 16 | 1741 | |||
GenBank mRNA | AB018290.1 | - | 15, 16 | 1528 | ||
AK022790.1 | - | 15, 16 | 1692 | |||
AK025463.1 | - | 11, 12 | 946 | |||
AK074368.1 | - | 4, 5 | 269 | |||
AK128730.1 | - | 14, 15 | 1545 | |||
AK225035.1 | - | 15, 16 | 1706 | |||
AK290337.1 | - | 15, 16 | 1676 | |||
AL050134.1 | - | 7, 8 | 489 | |||
BC004998.1 | - | 15, 16 | 1673 | 2958094 | ||
DQ993200.1 | - | 15, 16 | 1623 |