Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015306.2 | NP_056121.2 | 49 | 6023 | ||
XM_005270690.3 | XP_005270747.1 | 36 | 4064 | |||
XM_017000831.1 | XP_016856320.1 | 50 | 6088 | |||
XM_017000832.1 | XP_016856321.1 | 50 | 6085 | |||
XM_017000833.1 | XP_016856322.1 | 50 | 5580 | |||
XM_017000834.1 | XP_016856323.1 | 49 | 5520 | |||
GenBank mRNA | AB028980.1 | - | 8 | 842 | ||
AK127075.1 | - | 32 | 3768 | |||
AL832370.1 | - | 11 | 1249 |