Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QPRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318249.1 | NP_001305178.1 | 3 | 378 | ||
NM_001318250.1 | NP_001305179.1 | 4 | 470 | |||
NM_014298.4 | NP_055113.2 | 4 | 914 | |||
XM_005255223.3 | XP_005255280.2 | 5 | 1000 | |||
XM_017023101.1 | XP_016878590.1 | 5 | 997 | |||
GenBank mRNA | AK090801.1 | - | 1 | 1449 | ||
BC005060.1 | - | 4 | 751 | 2960170 | ||
BC010033.2 | - | 4 | 743 | 3506460 | ||
BC018910.2 | - | 4 | 768 | 3959973 | ||
BJ997057.1 | - | 2, 3 | 270 | |||
BP314322.1 | - | 4 | 296 | |||
BT007231.1 | - | 3 | 724 | |||
D78177.1 | - | 3 | 724 | |||
EU794638.1 | - | 4 | 743 | |||
KJ534923.1 | - | 3 | 724 |