Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015351.1 | NP_056166.1 | 1 | 544 | ||
GenBank mRNA | BC047950.1 | - | 1 | 366 | ||
BX407741.2 | - | 1 | 371 | |||
D86980.1 | - | 1 | 544 |