Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPIDR SIRNA Assays ›
Gene Symbol:
SPIDR
Gene Name:
scaffolding protein involved in DNA repair
Gene Aliases:
KIAA0146
Chromosome Location:
Chr.8: 47260878 - 47736001 on Build GRCh38
UniGene ID:
Hs.381058
Species:
Human
siRNA ID
HSS146453
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SPIDR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080394.3
NP_001073863.1
9
1219
NM_001282916.1
NP_001269845.1
8
1152
NM_001282919.1
NP_001269848.1
9
1182
NR_104581.1
7
867
XM_005251189.3
XP_005251246.1
9
1161
XM_005251191.3
XP_005251248.1
9
1161
XM_005251193.3
XP_005251250.1
9
1161
XM_005251195.4
XP_005251252.1
8
1005
XM_005251198.4
XP_005251255.1
8
939
XM_005251199.4
XP_005251256.1
6
753
XM_006716443.3
XP_006716506.1
9
1129
XM_006716444.3
XP_006716507.1
8
902
XM_011517497.2
XP_011515799.1
9
1161
XM_011517500.2
XP_011515802.1
9
1059
XM_017013268.1
XP_016868757.1
9
1161
XM_017013269.1
XP_016868758.1
9
1161
XM_017013270.1
XP_016868759.1
9
1059
XM_017013271.1
XP_016868760.1
9
1276
XM_017013272.1
XP_016868761.1
5
905
XM_017013273.1
XP_016868762.1
7
719
XM_017013274.1
XP_016868763.1
8
1005
XM_017013275.1
XP_016868764.1
5
757
XM_017013276.1
XP_016868765.1
5
744
XM_017013277.1
XP_016868766.1
9
1146
XM_017013278.1
XP_016868767.1
8
971
XM_017013279.1
XP_016868768.1
9
1149
XM_017013280.1
XP_016868769.1
8
939
XM_017013281.1
XP_016868770.1
8
904
XM_017013282.1
XP_016868771.1
8
1005
XM_017013283.1
XP_016868772.1
9
1149
XM_017013284.1
XP_016868773.1
8
1005
XM_017013285.1
XP_016868774.1
8
939
XM_017013286.1
XP_016868775.1
6
753
XM_017013287.1
XP_016868776.1
7
771
XM_017013288.1
XP_016868777.1
4
523
XM_017013289.1
XP_016868778.1
5
659
XM_017013290.1
XP_016868779.1
6
753
XM_017013291.1
XP_016868780.1
10
1184
GenBank mRNA
AK026929.1
-
2
183
AK122820.1
-
10
1517
AK293809.1
-
9
1142
AK293972.1
-
7
790
AK294248.1
-
5
580
AK294275.1
-
8
1074
AK297681.1
-
8
890
AK303579.1
-
9
1129
BC031627.1
-
7
792
D63480.1
-
9
1147
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
cellular response to DNA damage stimulus
regulation of double-strand break repair via homologous recombination
positive regulation of protein complex assembly
regulation of establishment of protein localization to chromosome
cellular response to ionizing radiation
cellular response to hydroxyurea
cellular response to camptothecin
positive regulation of double-strand break repair
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline