Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SOSTDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015464.2 | NP_056279.1 | 2 | 670 | ||
XM_017011943.1 | XP_016867432.1 | 3 | 812 | |||
GenBank mRNA | AB059270.1 | - | 2 | 489 | ||
AF361494.1 | - | 2 | 670 | |||
AK093408.1 | - | 5 | 1274 | |||
AK315682.1 | - | 2 | 557 | |||
AL050024.1 | - | 1 | 273 | |||
BC008484.1 | - | 2 | 559 | 4048654 |