Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMACHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015506.2 | NP_056321.2 | 2 | 312 | ||
XM_011541204.2 | XP_011539506.1 | 2 | 221 | |||
GenBank mRNA | AK098537.1 | - | 2 | 312 | ||
AL080062.1 | - | 2 | 183 | |||
BC006122.1 | - | 2 | 155 | 3535453 | ||
BU839693.1 | - | 2 | 167 |