Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NAT9 SIRNA Assays ›
Gene Symbol:
NAT9
Gene Name:
N-acetyltransferase 9 (putative)
Gene Aliases:
EBSP, hNATL
Chromosome Location:
Chr.17: 74770529 - 74776425 on Build GRCh38
UniGene ID:
Hs.144058
Species:
Human
siRNA ID
HSS146712
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NAT9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001305077.1
NP_001292006.1
7
746
NM_001305078.1
NP_001292007.1
7
745
NM_001305079.1
NP_001292008.1
7
672
NM_001305080.1
NP_001292009.1
7
734
NM_001305081.1
NP_001292010.1
7
743
NM_001305082.1
NP_001292011.1
7
894
NM_001305083.1
NP_001292012.1
7
731
NM_001305084.1
NP_001292013.1
6
630
NM_001305085.1
NP_001292014.1
6
627
NM_001305086.1
NP_001292015.1
6
651
NM_001305087.1
NP_001292016.1
6
650
NM_001305088.1
NP_001292017.1
6
583
NM_015654.4
NP_056469.2
7
749
NR_130953.1
8
753
NR_130954.1
7
631
XM_005257212.4
XP_005257269.1
6
588
XM_006721813.3
XP_006721876.1
6
1036
XM_017024437.1
XP_016879926.1
7
675
XM_017024439.1
XP_016879928.1
7
639
XM_017024441.1
XP_016879930.1
5
929
GenBank mRNA
AK123115.1
-
6
962
AK225602.1
-
7
619
AK308569.1
-
7
1027
AK312958.1
-
7
619
AL050269.1
-
7
626
AL537925.3
-
7
826
AY632082.1
-
6
585
BC004195.2
-
7
655
3528894
BC004225.2
-
7
644
3538186
BG702300.1
-
6
563
BI821280.1
-
8
650
BM464121.1
-
6
557
BM909550.1
-
7
633
BQ068039.1
-
6
465
BU159218.1
-
7
641
BX333236.2
-
6
564
DB171198.1
-
6
544
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
N-terminal protein amino acid acetylation
protein binding
N-acetyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0