Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080453.2 | NP_001073922.2 | 48 | 6644 | ||
XM_011515260.1 | XP_011513562.1 | 48 | 6674 | |||
XM_017011959.1 | XP_016867448.1 | 47 | 6827 | |||
XM_017011960.1 | XP_016867449.1 | 47 | 6797 | |||
GenBank mRNA | AB037861.1 | - | 30 | 4104 | ||
AL050110.1 | - | 15 | 1894 | |||
AL137358.1 | - | 8 | 797 | |||
AY358482.1 | - | 23 | 3105 | |||
BC004286.1 | - | 29 | 3817 | |||
BC013367.2 | - | 18 | 2400 | |||
BC018777.1 | - | 16 | 2067 | |||
BC069262.1 | - | 20 | 2674 |