Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012166.2 | NP_036298.2 | 3 | 973 | ||
XM_005251439.4 | XP_005251496.1 | 3 | 1062 | |||
XM_017014618.1 | XP_016870107.1 | 3 | 1179 | |||
XM_017014619.1 | XP_016870108.1 | 3 | 1290 | |||
GenBank mRNA | AB209610.1 | - | 2 | 561 | ||
AF176705.1 | - | 3 | 972 | |||
BC125125.1 | - | 3 | 1052 | 40119019 | ||
BC140785.1 | - | 2 | 944 | 9021653 | ||
BC171785.1 | - | 2 | 944 | 9054439 |