Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IL17C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013278.3 | NP_037410.1 | 3 | 582 | ||
GenBank mRNA | AF152099.1 | - | 3 | 582 | ||
AY358471.1 | - | 3 | 582 | |||
BC069152.1 | - | 2 | 533 | 7217012 | ||
BC101835.1 | - | 3 | 577 | 8069341 | ||
BC112196.1 | - | 3 | 577 | 8327664 |