Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014453.3 | NP_055268.1 | 3 | 378 | ||
NM_198426.2 | NP_940818.1 | 3 | 380 | |||
XM_005258746.2 | XP_005258803.1 | 3 | 878 | |||
XM_005258747.3 | XP_005258804.1 | 3 | 1001 | |||
GenBank mRNA | AK311974.1 | - | 3 | 266 | ||
BC002502.2 | - | 3 | 350 | 3051024 | ||
BG546479.1 | - | 3 | 374 | |||
BI457978.1 | - | 3 | 370 | |||
BP315111.1 | - | 3 | 378 | |||
BT007298.1 | - | 2 | 217 | |||
BU147101.1 | - | 1 | 25 | |||
CN305066.1 | - | 3 | 381 | |||
CR457002.1 | - | 2 | 217 |