Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014453.3 | NP_055268.1 | 6 | 808 | ||
NM_198426.2 | NP_940818.1 | 6 | 810 | |||
XM_005258746.2 | XP_005258803.1 | 5 | 1437 | |||
XM_005258747.3 | XP_005258804.1 | 5 | 1560 | |||
GenBank mRNA | AF042384.1 | - | 6 | 776 | ||
AK311974.1 | - | 6 | 696 | |||
AY364248.1 | - | 5 | 647 | |||
BC002502.2 | - | 6 | 780 | 3051024 | ||
BE856471.1 | - | 1 | 104 | |||
BT007298.1 | - | 5 | 647 | |||
BU147101.1 | - | 4 | 455 | |||
CR457002.1 | - | 5 | 647 |