Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014047.2 | NP_054766.1 | 3 | 229 | ||
GenBank mRNA | AF078852.1 | - | 3 | 203 | ||
AF086483.1 | - | 3 | 201 | |||
AK311855.1 | - | 3 | 194 | |||
BC015465.2 | - | 3 | 204 | 3914049 | ||
DA768829.1 | - | 3 | 230 |