Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014167.4 | NP_054886.2 | 1 | 122 | ||
GenBank mRNA | AF161477.1 | - | 1 | 96 | ||
AF213377.1 | - | 1 | 89 | |||
AK001156.1 | - | 1 | 98 | |||
AK225139.1 | - | 1 | 101 | |||
BC020647.1 | - | 1 | 85 | 4771954 | ||
BG538660.1 | - | 1 | 127 |