Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N6AMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013240.5 | NP_037372.4 | 2 | 229 | ||
NM_182749.4 | NP_877426.4 | 2 | 229 | |||
NR_047510.2 | 2 | 229 | ||||
GenBank mRNA | AF139682.1 | - | 2 | 230 | ||
AK225515.1 | - | 2 | 223 | |||
AK314100.1 | - | 2 | 221 | |||
BC011554.1 | - | 2 | 210 | 4640080 | ||
BP317563.1 | - | 2 | 226 |